missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone (stabilized with K2CO3) 98.0+%, TCI America™

Catalog No. C086725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C086725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C086725ML Supplier TCI America Supplier No. C086725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5891-21-4
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00001008
InChI Key XVRIEWDDMODMGA-UHFFFAOYSA-N
Synonym 5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one
PubChem CID 79993
IUPAC Name 5-chloropentan-2-one
SMILES CC(=O)CCCCl

Specifications

Boiling Point 106°C
Quantity 25 mL
UN Number 1224
Formula Weight 120.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-pentanone (stabilized with K2CO3)
RTECSNumber SA8143500
TSCA Yes