missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. C02175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02175G 5 g
C021725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C02175G Supplier TCI America Supplier No. C02175G
Only null left

Chemical Identifiers

CAS 6628-86-0
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.563
MDL Number MFCD00007289
InChI Key SWGPIDCNYAYXMJ-UHFFFAOYSA-N
Synonym 2-nitro-5-chlorobenzaldehyde, 5-chloro-2-nitro-benzaldehyde, benzaldehyde, 5-chloro-2-nitro, 4-chloro-2-formylnitrobenzene, pubchem8199, acmc-1cuhe, ksc497q2h, 5-chloro-2-nitro benzoaldehyde, 5-chloro-2-nitrobenzaldehyde, technical grade
PubChem CID 81123
IUPAC Name 5-chloro-2-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)[N+](=O)[O-]

Specifications

Melting Point 76°C
Color Yellow
Quantity 5 g
Formula Weight 185.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrobenzaldehyde
EINECSNumber (3)-1190
TSCA No