missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzimidazole 98.0+%, TCI America™

Catalog No. C07665G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C07665G Supplier TCI America Supplier No. C07665G

Chemical Identifiers

CAS 2818-69-1
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.608
MDL Number MFCD00014559
InChI Key NICFDLORAOTXMD-UHFFFAOYSA-N
PubChem CID 76063
IUPAC Name 6-chloro-2-methyl-1H-benzimidazole
SMILES CC1=NC2=C(N1)C=C(C=C2)Cl

Specifications

Melting Point 208°C
Quantity 5 g
Formula Weight 166.61
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylbenzimidazole