missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylaniline Hydrochloride 98.0+%, TCI America™

Catalog No. C074625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C074625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C074625G Supplier TCI America Supplier No. C074625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6259-42-3
Molecular Formula C7H9Cl2N
Molecular Weight (g/mol) 178.056
MDL Number MFCD00035487
InChI Key BMZGSMUCRXYUGB-UHFFFAOYSA-N
Synonym 2-Amino-4-chlorotoluene Hydrochloride, 5-Chloro-o-toluidine Hydrochloride
PubChem CID 80416
IUPAC Name 5-chloro-2-methylaniline;hydrochloride
SMILES CC1=C(C=C(C=C1)Cl)N.Cl

Specifications

Quantity 25 g
Formula Weight 178.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylaniline Hydrochloride
EINECSNumber (3)-0265
TSCA Yes