missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH2712714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2712714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2712714 Supplier Thermo Scientific Chemicals Supplier No. H2712714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89694-48-4 | Molecular Formula: C7H8BClO3 | Molecular Weight: 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89694-48-4
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD01318966
InChI Key FMBVAOHFMSQDGT-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxyphenyl boronic acid, 5-chloro-2-methoxybenzeneboronic acid, 5-chloro-2-methoxyphenyl boranediol, 2-methoxy-5-chlorophenylboronic acid, 4-chloroanisole-2-boronic acid, boronic acid, 5-chloro-2-methoxyphenyl, 5-chloro-2-methoxyphenylboronicacid, pubchem1782, 5-chloro-2-methoxy-phenyl boronic acid
PubChem CID 2735751
IUPAC Name (5-chloro-2-methoxyphenyl)boronic acid
SMILES COC1=C(C=C(Cl)C=C1)B(O)O

Specifications

Melting Point 134°C to 141°C
Quantity 25 g
Formula Weight 186.4
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only