missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-hydroxybenzophenone 98.0+%, TCI America™
Supplier: TCI America C018625G
Specifications
| 5-Chloro-2-hydroxybenzophenone | |
| 97°C | |
| MFCD00020134 | |
| OMWSZDODENFLSV-UHFFFAOYSA-N | |
| 2-benzoyl-4-chlorophenol | |
| 6799 | |
| ≥98.0% (GC) |
| 85-19-8 | |
| C13H9ClO2 | |
| 25 g | |
| OC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1 | |
| 232.66 | |
| 232.66 | |
| Crystalline Powder |
Chemical Identifiers
| 85-19-8 | |
| 232.66 | |
| OMWSZDODENFLSV-UHFFFAOYSA-N | |
| 2-benzoyl-4-chlorophenol |
| C13H9ClO2 | |
| MFCD00020134 | |
| 6799 | |
| OC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (9)-0356
TSCA : No