missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxybenzophenone 98.0+%, TCI America™

Catalog No. p-7124267
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01865G 5 g
C018625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01865G Supplier TCI America Supplier No. C01865G
Only null left

Chemical Identifiers

CAS 85-19-8
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.66
MDL Number MFCD00020134
InChI Key OMWSZDODENFLSV-UHFFFAOYSA-N
PubChem CID 6799
IUPAC Name 2-benzoyl-4-chlorophenol
SMILES OC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 97°C
Quantity 5 g
Formula Weight 232.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-hydroxybenzophenone
EINECSNumber (9)-0356
TSCA No