accessibility menu, dialog, popup

UserName

CAS RN 85-19-8

OHClO

CAS RN 85-19-8

IUPAC Name: 2-benzoyl-4-chlorophenol
Synonyms: benzophenone-7
Molecular Weight (g/mol): 232.66
Molecular Formula: C13H9ClO2
InChi Key: OMWSZDODENFLSV-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results