missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-hydroxy-4-methylbenzophenone 98.0+%, TCI America™
Supplier: TCI America C326025G
Specifications
| 5-Chloro-2-hydroxy-4-methylbenzophenone | |
| 142°C | |
| C14H11ClO2 | |
| 25 g | |
| CC1=CC(O)=C(C=C1Cl)C(=O)C1=CC=CC=C1 | |
| 246.69 | |
| 246.69 | |
| Crystalline Powder |
| 68751-90-6 | |
| Yellow | |
| MFCD00002325 | |
| IOGQACQRFGDUPT-UHFFFAOYSA-N | |
| 2-benzoyl-4-chloro-5-methylphenol | |
| 555814 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 68751-90-6 | |
| 246.69 | |
| IOGQACQRFGDUPT-UHFFFAOYSA-N | |
| 2-benzoyl-4-chloro-5-methylphenol |
| C14H11ClO2 | |
| MFCD00002325 | |
| 555814 | |
| CC1=CC(O)=C(C=C1Cl)C(=O)C1=CC=CC=C1 |
Safety and Handling
TSCA : No