missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxy-4-methylbenzophenone 98.0+%, TCI America™

Catalog No. p-7125907
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32605G 5 g
C326025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32605G Supplier TCI America Supplier No. C32605G

Chemical Identifiers

CAS 68751-90-6
Molecular Formula C14H11ClO2
Molecular Weight (g/mol) 246.69
MDL Number MFCD00002325
InChI Key IOGQACQRFGDUPT-UHFFFAOYSA-N
PubChem CID 555814
IUPAC Name 2-benzoyl-4-chloro-5-methylphenol
SMILES CC1=CC(O)=C(C=C1Cl)C(=O)C1=CC=CC=C1

Specifications

Melting Point 142°C
Color Yellow
Quantity 5 g
Formula Weight 246.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-hydroxy-4-methylbenzophenone