accessibility menu, dialog, popup

UserName

CH3OHClO

CAS RN 68751-90-6

IUPAC Name: 2-benzoyl-4-chloro-5-methylphenol
Synonyms: 2-benzoyl-4-chloro-5-methylphenol
Molecular Weight (g/mol): 246.69
Molecular Formula: C14H11ClO2
InChi Key: IOGQACQRFGDUPT-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C=C1Cl)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results