missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7125836
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31145G Supplier TCI America Supplier No. C31145G

Chemical Identifiers

CAS 96515-79-6
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD03094518
InChI Key WDTUCEMLUHTMCB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chlorobenzaldehyde, 3-chloro-6-fluoro benzaldehyde, 3-chloro-6-fluorobenzaldehyde, 5-chloro-2-fluoro-benzaldehyde, benzaldehyde, 5-chloro-2-fluoro, 2-fluoro-5-chloro-benzaldehyde, zlchem 333, pubchem1411, ksc486m0t, 2-fluoro-5-chloro benzaldehyde
PubChem CID 2773586
IUPAC Name 5-chloro-2-fluorobenzaldehyde
SMILES FC1=CC=C(Cl)C=C1C=O

Specifications

Melting Point 37°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-fluorobenzaldehyde