missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-benzothiazolinone 98.0+%, TCI America™

Catalog No. C113725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C113725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C113725G Supplier TCI America Supplier No. C113725G
Only null left

Chemical Identifiers

CAS 20600-44-6
Molecular Formula C7H4ClNOS
Molecular Weight (g/mol) 185.63
MDL Number MFCD00060125
InChI Key NOVHYVKPKWACML-UHFFFAOYSA-N
PubChem CID 603829
IUPAC Name 5-chloro-2,3-dihydro-1,3-benzothiazol-2-one
SMILES ClC1=CC=C2SC(=O)NC2=C1

Specifications

Melting Point 237°C
Quantity 25 g
Formula Weight 185.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-benzothiazolinone