missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-dimethyl-1H-pyrazole, 98%

Numéro de catalogue. p-7081663
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1173006 5 g
AAA1173014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1173006 Fournisseur Thermo Scientific Chemicals Code fournisseur A1173006
Il en reste null

CAS: 54454-10-3 | C5H7ClN2 | 130.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 54454-10-3
Molecular Formula C5H7ClN2
Molecular Weight (g/mol) 130.58
MDL Number MFCD00051650
InChI Key DDUSLFAWARYAPR-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
PubChem CID 521507
IUPAC Name 5-chloro-1,3-dimethylpyrazole
SMILES CN1N=C(C)C=C1Cl

Spécifications

Density 1.137
Boiling Point 153°C to 155°C
Refractive Index 1.486
Quantity 5 g
Beilstein 113451
Formula Weight 130.58
Percent Purity 98%
Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only