missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Bromouracil, 98%

Catalog No. AC226440250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226440250 25 g Glass bottle
AC226440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226440250 Supplier Thermo Scientific Chemicals Supplier No. 226440250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51-20-7 | C4H3BrN2O2 | 190.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-20-7
Molecular Formula C4H3BrN2O2
Molecular Weight (g/mol) 190.98
MDL Number MFCD00006017
InChI Key LQLQRFGHAALLLE-UHFFFAOYSA-N
Synonym 5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 5802
ChEBI CHEBI:20552
IUPAC Name 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES BrC1=CNC(=O)NC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 307.0°C to 310.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 24,318
Merck Index 15,145
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in ammonia water and other alkalines
Formula Weight 190.98
Percent Purity 98%
Chemical Name or Material 5-Bromouracil, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 200-084-0
RTECSNumber YQ9060000
TSCA TSCA

RUO – Research Use Only