missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrimidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH64041MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64041MD 250 mg
AAH6404106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64041MD Supplier Thermo Scientific Chemicals Supplier No. H64041MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37131-87-6 | Molecular Formula: C5H3BrN2O2 | Molecular Weight: 202.995 g/mol

5-Bromopyrimidine-2-carboxylic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyrimidine-2-carboxylic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 37131-87-6
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 202.995
MDL Number MFCD00496793
InChI Key XGPTXUYKEDPXCO-UHFFFAOYSA-N
Synonym 5-bromo-pyrimidine-2-carboxylic acid, 5-bromo-2-pyrimidinecarboxylic acid, 2-pyrimidinecarboxylic acid, 5-bromo, 5-bromopyrimidine-2-carboxylicacid, pubchem16922, acmc-1agpr, ksc222c2t, 5-bromopyrimidine-2-caboxylic acid, abbypharma ap-10-1676, 2-pyrimidinecarboxylicacid, 5-bromo
PubChem CID 16462970
IUPAC Name 5-bromopyrimidine-2-carboxylic acid
SMILES C1=C(C=NC(=N1)C(=O)O)Br

Specifications

Melting Point 193°C to 198°C
Quantity 250 mg
Beilstein 121432
Solubility Information Slightly soluble in water.
Formula Weight 203
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromopyrimidine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only