missing translation for 'onlineSavingsMsg'
Learn More

5-Bromonicotinic Acid 99.0+%, TCI America™

Catalog No. B181825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B181825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B181825G Supplier TCI America Supplier No. B181825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity building block reagent suitable for research applications.

Chemical Identifiers

CAS 20826-04-4
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00009783
InChI Key FQIUCPGDKPXSLL-UHFFFAOYSA-N
Synonym 5-bromonicotinic acid, 5-bromo nicotinic acid, 5-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo, nicotinic acid, 5-bromo, 5-bromo-nicotinic acid, 3-bromo-5-pyridine carboxylic acid, 3-bromo pyridine-5-carboxylic acid, pubchem1211, acmc-209fev
PubChem CID 88707
IUPAC Name 5-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(Br)=C1

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 25 g
Formula Weight 202.01
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromonicotinic Acid
RTECSNumber QT0868000
TSCA No