missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-méthyl-2-phényl-1,3-thiazole, ≥97%, Thermo Scientific™

Numéro de catalogue. CC29210DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC29210DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC29210DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC29210DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Spécifications

Nom ou substance chimique 5-Bromo-4-methyl-2-phenyl-1, 3-thiazole
CAS 28771-82-6
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Formule moléculaire C10H8BrNS
Quantité 1 g
Numéro MDL MFCD04974049
Synonyme 5-bromo-4-methyl-2-phenylthiazole, thiazole,5-bromo-4-methyl-2-phenyl, 5-bromo-4-methyl-2-phenyl-thiazole, thiazole, 5-bromo-4-methyl-2-phenyl, 5-bromo-4-methyl-1,3-thiazol-2-yl benzene
Clé InChI XNYKJKJMPCGEQI-UHFFFAOYSA-N
SOURIRES CC1=C(Br)SC(=N1)C1=CC=CC=C1
Nom de l’IUPAC 5-bromo-4-méthyl-2-phényl-1,3-thiazole
Poids moléculaire (g/mol) 254.15
PubChem CID 2795494
Poids de formule 254.15
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.