missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolylphosphate Disodium Salt, ≥98%, MP Biomedicals™

Catalog No. ICN15004283
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15004283 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15004283 Supplier MP Biomedicals Inc Supplier No. 0215004283

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disodium salt is soluble in water (50mg/mL); insoluble in dimethylformamide

Chemical Identifiers

CAS 102185-33-1
Molecular Formula C8H4BrClNNa2O4P
Molecular Weight (g/mol) 370.43
MDL Number MFCD00036757
InChI Key OAZUOCJOEUNDEK-UHFFFAOYSA-L
Synonym sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip™, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
PubChem CID 6097197
IUPAC Name disodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Specifications

Color White
Assay Percent Range ≥98%
Quantity 250 mg
Formula Weight 370.4
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolylphosphate Disodium Salt