missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt, 98%

Catalog No. AAJ64360MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64360MD 250 mg
AAJ6436003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64360MD Supplier Thermo Scientific Chemicals Supplier No. J64360MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for beta-glucuronidase | CAS: 129541-41-9 | C14H12BrClNNaO7 | 444.594 g/mol

5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt is a chromogenic substrate for β-glucuronidase (GUS) gene detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt is a chromogenic substrate for β-glucuronidase (GUS) gene detection.

Solubility
Soluble in water at 10mg/ml

Notes
Light sensitive. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 129541-41-9
Molecular Formula C14H12BrClNNaO7
Molecular Weight (g/mol) 444.594
MDL Number MFCD00135782
InChI Key IBLSVGDGSKUDCT-ILIJQVQCSA-M
Synonym x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
PubChem CID 53384407
IUPAC Name sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]

Specifications

Color White
Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Soluble in water at 10mg/ml
Formula Weight 444.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only