missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridinecarboxaldéhyde 98,0+%, TCI America™

Catalog No. B34905G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B34905G 5 g
B34901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34905G Supplier TCI America Supplier No. B34905G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 113118-81-3
Formule moléculaire C6H4BrNO
Poids moléculaire (g/mol) 186.008
Numéro MDL MFCD03265758
Clé InChI NGUVGKAEOFPLDT-UHFFFAOYSA-N
Synonyme 5-bromonicotinaldehyde, 5-bromo-3-pyridinecarboxaldehyde, 5-bromo-3-formylpyridine, 3-bromopyridine-5-carboxaldehyde, 5-bromopyridine-3-carboxaldehyde, 5-bromo-pyridine-3-carbaldehyde, 3-bromo-5-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo, 3-bromopyridine-5-carbaldehyde, 5-bromo-3-pyridinecarbaldehyde
PubChem CID 2784734
Nom de l’IUPAC 5-bromopyridine-3-carbaldehyde
SOURIRES C1=C(C=NC=C1Br)C=O

Specifications

Nom ou substance chimique 5-Bromo-3-pyridinecarboxaldehyde
Point de fusion 98 °C
Quantité 5 g
Poids de formule 186.01
Pourcentage de pureté ≥98,0% (GC)
Forme physique Poudre cristalline