missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. B34511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34511G 1 g
B34515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34511G Supplier TCI America Supplier No. B34511G

Chemical Identifiers

CAS 436799-32-5
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD06657686
InChI Key RPFAUCIXZGMCFN-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
PubChem CID 2761197
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1Br)C(F)(F)F

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 78°C
Quantity 1 g
UN Number 2811
Formula Weight 226.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine