missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH3231603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3231603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3231603 Supplier Thermo Scientific Chemicals Supplier No. H3231603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 749875-16-9 | Molecular Formula: C7H3BrF3NO2 | Molecular Weight: 270.01 g/mol

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 749875-16-9
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.01
MDL Number MFCD10687236
InChI Key XNVAVQMVAYUTBA-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethyl isonicotinic acid, 5-bromo-2-trifluoromethyl pyridine-4-carboxylic acid, 5-bromo-2-trifluoromethyl-isonicotinic acid, 4-pyridinecarboxylic acid, 5-bromo-2-trifluoromethyl, 5-bromo-2-trifluoromethyl isonicotinicacid, 3-bromo-6-trifluoromethyl pyridine-4-carboxylic acid
PubChem CID 28306936
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC=C1Br)C(F)(F)F

Specifications

Melting Point 235°C to 238°C
Quantity 1 g
Solubility Information Slightly soluble in water,soluble in organic solvents.
Formula Weight 270
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only