missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thiophenecarboxaldehyde, 97%

Catalog No. AC166651000
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166651000 100 mL Glass bottle
AC166650050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166651000 Supplier Thermo Scientific Chemicals Supplier No. 166651000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4701-17-1 | C5H3BrOS | 191.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4701-17-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005432
InChI Key GFBVUFQNHLUCPX-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
PubChem CID 78428
IUPAC Name 5-bromothiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.6100g/mL
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6380 to 1.641
Quantity 100 mL
Beilstein 17, I, 148
Specific Gravity 1.61
Formula Weight 191.05
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 5-Bromo-2-thiophenecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-176-8

RUO – Research Use Only