missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylphenol, 95%

Catalog No. AAH2695603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2695603 1 g
AAH2695609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2695603 Supplier Thermo Scientific Chemicals Supplier No. H2695603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 36138-76-8 | C7H7BrO | 187.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36138-76-8
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD08061906
InChI Key OONJCAWRVJDVBB-UHFFFAOYSA-N
Synonym 5-bromo-o-cresol, phenol, 5-bromo-2-methyl, 4-bromo-2-hydroxytoluene, 5-bromo-2-methyl phenol, 5-bromo-2-methyl-phenol, 3-bromo-6-methylphenol, 2-hydroxy-4-bromotoluene, 5-brom-2-kresol, zlchem 468, pubchem4132
PubChem CID 12648404
IUPAC Name 5-bromo-2-methylphenol
SMILES CC1=C(C=C(C=C1)Br)O

Specifications

Melting Point 77°C to 80°C
Quantity 1 g
Formula Weight 187.04
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-methylphenol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only