missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzonitrile 98.0+%, TCI America™

Catalog No. B31235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31235G 5 g
B312325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31235G Supplier TCI America Supplier No. B31235G

Chemical Identifiers

CAS 144649-99-0
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD00143429
InChI Key LOASAXVECBZCRJ-UHFFFAOYSA-N
PubChem CID 4418560
IUPAC Name 5-bromo-2-methoxybenzonitrile
SMILES COC1=C(C=C(Br)C=C1)C#N

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Synonym 5-bromo-2-methoxy-benzonitrile, 4-bromo-2-cyanoanisole, benzonitrile,5-bromo-2-methoxy, 5-bromo-2-methyloxy benzonitrile, benzonitrile, 5-bromo-2-methoxy, 5-bromo-2-methoxybenzenecarbonitrile, acmc-1c8aq, timtec-bb sbb020096, art-chem-bb b005860, 5-bromo-2-methoxybenzonitrile
Formula Weight 212.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxybenzonitrile