missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydrazinopyridine 96.0+%, TCI America™

Catalog No. B43275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43275G 5 g
B43271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43275G Supplier TCI America Supplier No. B43275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77992-44-0
Molecular Formula C5H6BrN3
Molecular Weight (g/mol) 188.028
MDL Number MFCD00513424
InChI Key QYQLEYTXFMOLEI-UHFFFAOYSA-N
Synonym (5-Bromo-2-pyridyl)hydrazine
PubChem CID 11535699
IUPAC Name (5-bromopyridin-2-yl)hydrazine
SMILES C1=CC(=NC=C1Br)NN

Specifications

Melting Point 134°C
Color Yellow
Quantity 5 g
Formula Weight 188.03
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydrazinopyridine