missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-furoic Acid 98.0+%, TCI America™

Catalog No. B100825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B100825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B100825G Supplier TCI America Supplier No. B100825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 585-70-6
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00003239
InChI Key YVTQHZDUDUCGRD-UHFFFAOYSA-N
Synonym 5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
PubChem CID 68511
IUPAC Name 5-bromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)Br)C(=O)O

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 25 g
Formula Weight 190.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-furoic Acid