missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoropyridine 98.0+%, TCI America™

Catalog No. B27055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27055G 5 g
B270525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27055G Supplier TCI America Supplier No. B27055G

Chemical Identifiers

CAS 766-11-0
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
InChI Key MYUQKYGWKHTRPG-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5
PubChem CID 2783168
IUPAC Name 5-bromo-2-fluoropyridine
SMILES C1=CC(=NC=C1Br)F

Specifications

Color Yellow
Boiling Point 63°C
Quantity 5 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluoropyridine