missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoropyridine, 99%

Catalog No. p-3212208
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378460050 5 g Glass bottle
AC378460250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378460050 Supplier Thermo Scientific Chemicals Supplier No. 378460050

CAS: 766-11-0 | C5H3BrFN | 175.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-11-0
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.99
MDL Number MFCD01863742
InChI Key MYUQKYGWKHTRPG-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5
PubChem CID 2783168
IUPAC Name 5-bromo-2-fluoropyridine
SMILES C1=CC(=NC=C1Br)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.7200g/mL
Boiling Point 170.0°C to 173.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5310 to 1.5330
Quantity 5 g
Specific Gravity 1.72
Formula Weight 175.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluoropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia

RUO – Research Use Only