missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzaldehyde 95.0+%, TCI America™

Catalog No. B17895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17895G 5 g
B178925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17895G Supplier TCI America Supplier No. B17895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93777-26-5
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00070755
InChI Key MMFGGDVQLQQQRX-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
PubChem CID 736327
IUPAC Name 5-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 203.01
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Bromo-2-fluorobenzaldehyde