missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzaldehyde, 98%

Catalog No. AAA1421930
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1421930 250 g
AAA1421909 10 g
AAA1421918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1421930 Supplier Thermo Scientific Chemicals Supplier No. A1421930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 93777-26-5 | C7H4BrFO | 203.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93777-26-5
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00070755
InChI Key MMFGGDVQLQQQRX-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
PubChem CID 736327
IUPAC Name 5-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)F

Specifications

Density 1.71
Boiling Point 230°C
Flash Point 109°C (228°F)
Refractive Index 1.57
Quantity 250 g
Beilstein 7632798
Sensitivity Air Sensitive
Formula Weight 203.01
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 298-056-6
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only