missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoro-m-xylene, 97%

Catalog No. AAH5056406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5056406 5 g
AAH5056403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5056406 Supplier Thermo Scientific Chemicals Supplier No. H5056406

CAS: 99725-44-7 | C8H8BrF | 203.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99725-44-7
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD01320701
InChI Key ZXPHUVHMBKRRJF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-m-xylene, 5-bromo-2-fluoro-3-xylene, 4-bromo-2,6-dimethylfluorobenzene, 1-bromo-4-fluoro-3,5-dimethylbenzene, 5-bromo-2-fluoro-1,3-xylene, benzene, 5-bromo-2-fluoro-1,3-dimethyl, 3,5-dimethyl-4-fluorobromobenzene, 5-bromo-2-fluoro-1,3-dimethyl-benzene, ksc486i9j
PubChem CID 2736298
IUPAC Name 5-bromo-2-fluoro-1,3-dimethylbenzene
SMILES CC1=CC(=CC(=C1F)C)Br

Specifications

Quantity 5 g
Boiling Point 95°C (22 mmHg)
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-fluoro-m-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only