missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(difluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6629703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6629703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6629703 Supplier Thermo Scientific Chemicals Supplier No. H6629703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 845827-13-6 | Molecular Formula: C6H4BrF2N | Molecular Weight: 208.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 845827-13-6
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD11977429
InChI Key QXLZRIGSWWQOLG-UHFFFAOYSA-N
Synonym 5-bromo-2-difluoromethyl pyridine, pyridine, 5-bromo-2-difluoromethyl, 2-difluoromethyl-5-bromopyridine, pyridine,5-bromo-2-difluoromethyl, 3-bromo-6-difluoromethyl pyridine, 2-bis fluoranyl methyl-5-bromanyl-pyridine, 5-bromo-alpha,alpha-difluoro-2-picoline
PubChem CID 53415062
IUPAC Name 5-bromo-2-(difluoromethyl)pyridine
SMILES FC(F)C1=NC=C(Br)C=C1

Specifications

Melting Point 35°C to 40°C
Color White to Yellow
Flash Point 89°C (192°F)
Quantity 1 g
UN Number UN2811
Formula Weight 208
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-(difluoromethyl)pyridine
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only