missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals
Description
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a pharmaceutical intermediate.
Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol
Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 124°C to 127°C |
| Quantity | 5 g |
| Beilstein | 5498624 |
| Solubility Information | Soluble in dichloromethane,ether,ethyl acetate and methanol |
| Formula Weight | 183.01 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Bromo-2-cyanopyridine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only