missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroanisole, 98%, Thermo Scientific™

Numéro de catalogue. AC398900050
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC398900050 5g Glass bottle
AC398900250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC398900050 Fournisseur Thermo Scientific Chemicals Code fournisseur 398900050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 16817-43-9
Formule moléculaire C7H6BrClO
Poids moléculaire (g/mol) 221.48
Numéro MDL MFCD00078659
Clé InChI UAMVKOTWSHJOSY-UHFFFAOYSA-N
Synonyme 5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
PubChem CID 2769567
Nom de l’IUPAC 4-bromo-1-chloro-2-methoxybenzene
SOURIRES COC1=C(Cl)C=CC(Br)=C1

Spécifications

Nom ou substance chimique 5-Bromo-2-chloroanisole
CAS % minimum 97.5
CAS % maximal 100.0
Densité 1.6310g/mL
Point d’ébullition 240°C to 245°C (748.0mmHg)
Point d’éclair 113°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97.5% min. (GC)
Emballage Glass bottle
Formule linéaire BrC6H3(Cl)OCH3
Quantité 5g
Gravité spécifique 1.631
Poids de formule 221.48
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only