missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-3-nitropyridine 98.0+%, TCI America™

Catalog No. B39635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39635G 5 g
B396325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39635G Supplier TCI America Supplier No. B39635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67443-38-3
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.44
MDL Number MFCD00222270
InChI Key WWQQPSDIIVXFOX-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
PubChem CID 7019412
IUPAC Name 5-bromo-2-chloro-3-nitropyridine
SMILES [O-][N+](=O)C1=CC(Br)=CN=C1Cl

Specifications

Melting Point 68°C
Color Yellow
Quantity 5 g
Formula Weight 237.44
Percent Purity ≥98.0% (GC)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloro-3-nitropyridine