missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-3-cyanopyridine 98.0+%, TCI America™

Catalog No. B48705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48705G 5 g
B48701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48705G Supplier TCI America Supplier No. B48705G

Chemical Identifiers

CAS 405224-23-9
Molecular Formula C6H2BrClN2
Molecular Weight (g/mol) 217.45
MDL Number MFCD09801046
InChI Key MQOHJAYYYVQBSH-UHFFFAOYSA-N
Synonym 5-bromo-2-chloronicotinonitrile, 5-bromo-2-chloro-3-cyanopyridine, 5-bromo-2-chloro-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 5-bromo-2-chloro, pubchem15627, acmc-1amn3, 5-bromo-2-chloronicotinontrile, 5-bromo-2-chloro-nicotinonitrile, 5-bromo-2-chloro-pyridine-3-carbonitrile, 5-bromo-2-chloropyridine-3-carbonitrile
PubChem CID 21948269
IUPAC Name 5-bromo-2-chloropyridine-3-carbonitrile
SMILES ClC1=NC=C(Br)C=C1C#N

Specifications

Melting Point 140°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 217.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloro-3-cyanopyridine