missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-diphenyl-1,3-thiazole, ≥97%, Thermo Scientific™

Numéro de catalogue. CC42310DA
Modifier l'affichage
Click to view available options
Quantité:
1g
Emballage:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC42310DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC42310DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC42310DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 35342-50-8
Formule moléculaire C15H10BrNS
Poids moléculaire (g/mol) 316.22
Numéro MDL MFCD08060523
Clé InChI KKDPGKYZSJFNLZ-UHFFFAOYSA-N
Synonyme 5-bromo-2,4-diphenylthiazole, thiazole,5-bromo-2,4-diphenyl, thiazole, 5-bromo-2,4-diphenyl
PubChem CID 7537557
Nom de l’IUPAC 5-bromo-2,4-diphenyl-1,3-thiazole
SOURIRES BrC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Spécifications

Nom ou substance chimique 5-Bromo-2, 4-diphenyl-1, 3-thiazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 316.22
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.