missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,2-difluoro-1,3-benzodioxole 97.0+%, TCI America™

Catalog No. B37815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37815G 5 g
B378125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37815G Supplier TCI America Supplier No. B37815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33070-32-5
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD00236212
InChI Key SZRHWHHXVXSGMT-UHFFFAOYSA-N
Synonym 5-bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2h-1,3-benzodioxole, 5-bromo-2,2-difluorobenzo d 1,3 dioxole, 5-bromo-2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo-2,2-difluoro, 4-bromo-1,2-difluoromethylenedioxy benzene, 5-bromo-2,2-difluorobenzo d 1,3-dioxolene, acmc-209hy8, 5-bromo2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-5-bromo-1,3-benzodioxole
PubChem CID 2736271
IUPAC Name 5-bromo-2,2-difluoro-1,3-benzodioxole
SMILES C1=CC2=C(C=C1Br)OC(O2)(F)F

Specifications

Color Colorless
Boiling Point 79°C
Quantity 5 g
Formula Weight 237.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2,2-difluoro-1,3-benzodioxole