missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-[2,2'-bithiophene]-5-carbonitrile 98.0+%, TCI America™

Catalog No. B5457200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5457200MG 200 mg
B54571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5457200MG Supplier TCI America Supplier No. B5457200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 176787-96-5
Molecular Formula C9H4BrNS2
Molecular Weight (g/mol) 270.16
MDL Number MFCD16339405
InChI Key OHVCHDZJRUEOKH-UHFFFAOYSA-N
PubChem CID 15458991
IUPAC Name 5'-bromo-[2,2'-bithiophene]-5-carbonitrile
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C#N

Specifications

Melting Point 163°C
Quantity 200 mg
Formula Weight 270.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Bromo-[2,2′-bithiophene]-5-carbonitrile