missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-benzofuran, 97%, Thermo Scientific™

Chemical Identifiers

CAS 23145-07-5
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
MDL Number MFCD03407317
InChI Key AYOVPQORFBWFNO-UHFFFAOYSA-N
Synonym 5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
PubChem CID 90015
IUPAC Name 5-bromo-1-benzofuran
SMILES C1=CC2=C(C=CO2)C=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC07110CB
SDS View Documents
Thermo Scientific Chemicals
CC07110CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC07110DE
SDS View Documents
Thermo Scientific Chemicals
CC07110DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23145-07-5
197.031
AYOVPQORFBWFNO-UHFFFAOYSA-N
90015
C1=CC2=C(C=CO2)C=C1Br
C8H5BrO
MFCD03407317
5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
5-bromo-1-benzofuran

Specifications

23145-07-5
100.0
Amber Glass Bottle
MFCD03407317
5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
C1=CC2=C(C=CO2)C=C1Br
197.031
197.03
5-Bromo-1-benzofuran
96.5
97%
C8H5BrO
250mg
AYOVPQORFBWFNO-UHFFFAOYSA-N
5-bromo-1-benzofuran
90015
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'