missing translation for 'onlineSavingsMsg'
Learn More

(5-Bromo-1-benzofuran-2-yl)methanol, ≥97%, Thermo Scientific™

Numéro de catalogue. CC31209CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC31209CB 250mg
CC31209DA 1g
CC31209DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC31209CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC31209CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 38220-77-8
Formule moléculaire C9H7BrO2
Poids moléculaire (g/mol) 227.06
Numéro MDL MFCD06659073
Clé InChI JYYWIDBNICYLBN-UHFFFAOYSA-N
Synonyme 5-bromo-1-benzofuran-2-yl methanol, 5-bromobenzofuran-2-yl methanol, 5-bromobenzo d furan-2-yl methan-1-ol, 5-bromo-2-benzofuranylmethanol, 5-bromo-2-benzofuranyl methanol, 5-bromo-benzofuran-2-yl-methanol, 5-bromo-2-hydroxymethyl-1-benzofuran, 5-bromo-1-benzo b furan-2-yl methanol
PubChem CID 2795537
Nom de l’IUPAC (5-bromo-1-benzofuran-2-yl)methanol
SOURIRES OCC1=CC2=CC(Br)=CC=C2O1

Spécifications

Nom ou substance chimique (5-Bromo-1-benzofuran-2-yl)methanol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 227.06
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.