missing translation for 'onlineSavingsMsg'
Learn More

5-(Boc-amino)pyridine-3-boronic acid pinacol ester, 95%

Catalog No. AAH66223MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66223MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66223MD Supplier Thermo Scientific Chemicals Supplier No. H66223MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1171897-39-4 | C16H25BN2O4 | 320.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1171897-39-4
Molecular Formula C16H25BN2O4
Molecular Weight (g/mol) 320.20
MDL Number MFCD13195277
InChI Key BYXPGPHNPNIEPY-UHFFFAOYSA-N
Synonym tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl carbamate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-ylcarbamate, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-3-yl-carbamic acid tert-butyl ester, 3-boc-aminopyridine-5-boronic acid pinacol ester, n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-3-yl-carbamic acid tert-butyl ester, 5-boc-amino pyridine-3-boronic acid pinacol ester, 5-n-boc-amino-pyridin-3-ylboronic acid pinacol ester, n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl pivalamide
PubChem CID 57416500
IUPAC Name tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CN=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Beilstein 19330771
Formula Weight 320.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-(Boc-amino)pyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only