missing translation for 'onlineSavingsMsg'
Learn More

5-(Boc-amino)-2-chlorobenzoic acid, 98+%

Catalog No. AAH5230806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5230806 5 g
AAH5230803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5230806 Supplier Thermo Scientific Chemicals Supplier No. H5230806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 503555-96-2 | C12H14ClNO4 | 271.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 503555-96-2
Molecular Formula C12H14ClNO4
Molecular Weight (g/mol) 271.70
MDL Number MFCD02682166
InChI Key QNRXWUBPCIBQMX-UHFFFAOYSA-N
Synonym 5-tert-butoxycarbonyl amino-2-chlorobenzoic acid, n-boc-5-amino-2-chlorobenzoic acid, 5-boc-amino-2-chlorobenzoic acid, 5-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-5-2-methyl-2-propanyl oxy carbonyl amino benzoic acid, benzoic acid, 2-chloro-5-1,1-dimethylethoxy carbonyl amino-9ci
PubChem CID 22291441
IUPAC Name 2-chloro-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC(C(O)=O)=C(Cl)C=C1

Specifications

Quantity 5 g
Formula Weight 271.7
Percent Purity ≥98%
Chemical Name or Material 5-(Boc-amino)-2-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only