missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyl L-Glutamate 98.0+%, TCI America™

Catalog No. B33165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33165G Supplier TCI America Supplier No. B33165G

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 158°C
Color White
Quantity 5 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyl L-Glutamate