missing translation for 'onlineSavingsMsg'
Learn More

5-Benzotriazolecarboxylic Acid 96.0+%, TCI America™

Catalog No. B38695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38695G 5 g
B386925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38695G Supplier TCI America Supplier No. B38695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23814-12-2
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00012318
InChI Key GUOVBFFLXKJFEE-UHFFFAOYSA-N
Synonym 5-Carboxybenzotriazole
PubChem CID 72917
IUPAC Name 2H-benzotriazole-5-carboxylic acid
SMILES C1=CC2=NNN=C2C=C1C(=O)O

Specifications

Quantity 5 g
Formula Weight 163.14
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzotriazolecarboxylic Acid
EINECSNumber (5)-5993&(5)-5794
TSCA No