missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azoniaspiro[4.4]nonane Iodide 98.0+%, TCI America™
Supplier: TCI America A30931G
Specifications
| 5-Azoniaspiro[4.4]nonane Iodide | |
| White | |
| 1 g | |
| C1CC[N+]2(C1)CCCC2.[I-] | |
| 253.127 | |
| 253.13 | |
| Crystalline Powder |
| 45650-35-9 | |
| C8H16IN | |
| DWOWCUCDJIERQX-UHFFFAOYSA-M | |
| 5-azoniaspiro[4.4]nonane;iodide | |
| 86209376 | |
| ≥98.0% (T) |
Chemical Identifiers
| 45650-35-9 | |
| 253.127 | |
| 86209376 | |
| C1CC[N+]2(C1)CCCC2.[I-] |
| C8H16IN | |
| DWOWCUCDJIERQX-UHFFFAOYSA-M | |
| 5-azoniaspiro[4.4]nonane;iodide |
Safety and Handling
TSCA : No