missing translation for 'onlineSavingsMsg'
Learn More

5-Azoniaspiro[4.4]nonane Chloride 98.0+%, TCI America™

Catalog No. A30925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30925G 5 g
A30921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30925G Supplier TCI America Supplier No. A30925G

Chemical Identifiers

CAS 98997-63-8
Molecular Formula C8H16ClN
Molecular Weight (g/mol) 161.673
InChI Key HGAIUEUNFDXNNI-UHFFFAOYSA-M
PubChem CID 86139055
IUPAC Name 5-azoniaspiro[4.4]nonane;chloride
SMILES C1CC[N+]2(C1)CCCC2.[Cl-]

Specifications

Melting Point 295°C
Color White
Quantity 5 g
Formula Weight 161.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azoniaspiro[4.4]nonane Chloride