missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azacytidine 98.0+%, TCI America™
$62.66 - $326.56
Chemical Identifiers
| CAS | 320-67-2 |
|---|---|
| Molecular Formula | C8H12N4O5 |
| Molecular Weight (g/mol) | 244.21 |
| MDL Number | MFCD00006539 |
| InChI Key | NMUSYJAQQFHJEW-KVTDHHQDSA-N |
| Synonym | 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin |
| PubChem CID | 9444 |
| ChEBI | CHEBI:2038 |
| IUPAC Name | 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one |
| SMILES | NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
A2033100MG
|
N/A | 100 mg |
Each for $62.66
|
|
|||||
|
A20331G
|
N/A | 1 g |
Each for $326.56
|
|
|||||
Chemical Identifiers
| 320-67-2 | |
| 244.21 | |
| NMUSYJAQQFHJEW-KVTDHHQDSA-N | |
| 9444 | |
| 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one |
| C8H12N4O5 | |
| MFCD00006539 | |
| 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin | |
| CHEBI:2038 | |
| NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O |
Specifications
| 320-67-2 | |
| White | |
| MFCD00006539 | |
| NMUSYJAQQFHJEW-KVTDHHQDSA-N | |
| 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one | |
| 9444 | |
| 244.21 | |
| Crystalline Powder |
| 216°C | |
| C8H12N4O5 | |
| 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin | |
| NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O | |
| 244.21 | |
| CHEBI:2038 | |
| ≥98.0% (T) | |
| 5-Azacytidine |
Safety and Handling
RTECSNumber : XZ3017500
TSCA : No
Recommended Storage : Freezer